L(-)-Carnitine inner salt ≥99%
Supplier: BeanTown Chemical
Synonyms:
Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
215415-10G
215415-250G
215415-50G
BT215415-10GEA
79
USD
BT215415-10G
BT215415-250G
BT215415-50G
L(-)-Carnitine inner salt ≥99%
L(-)-Carnitine
CAS: 541-15-1; EC No: 208-768-0; MDL No: MFCD00038747; RTECS: BP2980000
Crystalline; Molecular Formula: C7H15NO3; MW: 161.2
Melting Point: 197-212°
- RTECS: BP2980000
- Crystalline
Formula:
C₇H₁₅NO₃ Melting Pt: 197…212 °C |
MDL Number:
MFCD00038747 CAS Number: 541-15-1 EINECS: 208-768-0 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...