2-Methylquinoline-6-carboxylic acid ≥97%

Supplier: BeanTown Chemical

215330-1G 215330-25G 215330-5G
BT215330-1GEA 26 USD
BT215330-1G BT215330-25G BT215330-5G
2-Methylquinoline-6-carboxylic acid ≥97%
2-Methylquinoline-6-carboxylic acid

CAS: 635-80-3; MDL No: MFCD00014566 Solid; Molecular Formula: C11H9NO2 ; MW: 187.19


  • Solid

Formula: C₁₁H₉NO₂
MW: 187.19 g/mol
MDL Number: MFCD00748504
CAS Number: 635-80-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR