6-Fluoroindole ≥98%
Supplier: BeanTown Chemical
Synonyms:
6-Fluoro-1H-indole
215325-1G
215325-25G
215325-5G
BT215325-1GEA
96
USD
BT215325-1G
BT215325-25G
BT215325-5G
6-Fluoroindole ≥98%
6-Fluoroindole
CAS: 399-51-9; MDL No: MFCD00056933
Crystalline; Molecular Formula: C8H6FN; MW: 135.14
Melting Point: 72-76°
Light Sensitive
- Light Sensitive
- Crystalline
Formula:
C₈H₆FN Melting Pt: 72…76 °C |
MDL Number:
MFCD00056933 CAS Number: 399-51-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...