2-Chloro-4-formylpyridine ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Chloropyridine-4-carboxaldehyde, 2-Chloroisonicotinaldehyde

215315-1G 215315-5G
BT215315-1GEA 205 USD
BT215315-1G BT215315-5G
2-Chloro-4-formylpyridine ≥97%
2-Chloro-4-formylpyridine

CAS: 101066-61-9; MDL No: MFCD06651557 Solid; Molecular Formula: C6H4ClNO; MW: 141.56 Melting Point: 46-50° Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 46…50 °C
MDL Number: MFCD06651557
CAS Number: 101066-61-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR