Calcium hydrogen orthophosphate dihydrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Calcium phosphate dibasic dihydrate, Calcium hydrogen phosphate dihydrate, Calcium hydrogenphosphate dihydrate, Dibasic Calcium Phosphate Dihydrate
215280-1KG
215280-5KG
BT215280-1KGEA
88.48
USD
BT215280-1KG
BT215280-5KG
Calcium hydrogen orthophosphate dihydrate ≥98%
Calcium hydrogen orthophosphate dihydrate
CAS: 7789-77-7; EC No: 231-826-1; MDL No: MFCD00149621
Powder; Linear Formula: CaHPO4·2H2O; MW: 172.09
Density (g/mL): 2.31
- Powder
Formula:
HCaO₄P·2H₂O MW: 172.09 g/mol Density: 2.31 g/mL |
MDL Number:
MFCD00149621 CAS Number: 7789-77-7 EINECS: 231-826-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...