2-Pyridinethiol ≥98%
Supplier: BeanTown Chemical
Synonyms:
Pyridine-2-thiol, 2-Mercaptopyridine
215255-100G
215255-25G
215255-5G
BT215255-100GEA
255.67
USD
BT215255-100G
BT215255-25G
BT215255-5G
2-Pyridinethiol ≥98%
2-Pyridinethiol
CAS: 2637-34-5; EC No: 220-131-9; MDL No: MFCD00006285; RTECS: UT9600000
Powder; Molecular Formula: C5H5NS; MW: 111.16
Melting Point: 127-130°
Air Sensitive
- RTECS: UT9600000
- Air Sensitive
- Powder
Formula:
C₅H₅NS MW: 111.17 g/mol Melting Pt: 127…130 °C |
MDL Number:
MFCD00006285 CAS Number: 2637-34-5 EINECS: 220-131-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...