Lithium trifluoromethanesulfonate ≥97%
Supplier: BeanTown Chemical
215240-10G
215240-50G
BT215240-10GEA
44.88
USD
BT215240-10G
BT215240-50G
Lithium trifluoromethanesulfonate ≥97%
Lithium trifluoromethanesulphonate
CAS: 33454-82-9; EC No: 251-528-5; MDL No: MFCD00013227
Powder; Linear Formula: LiCF3SO3 ; MW: 156.01
Melting Point:
- Moisture Sensitive
- Powder
Formula:
CF₃LiO₃S Melting Pt: >300 °C |
MDL Number:
MFCD00013227 CAS Number: 33454-82-9 EINECS: 251-528-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...