Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride ≥97%
Supplier: BeanTown Chemical
215225-100G
215225-25G
215225-5G
BT215225-100GEA
742
USD
BT215225-100G
BT215225-25G
BT215225-5G
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride ≥97%
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride
CAS: 4644-61-5; EC No: 225-073-8; MDL No: MFCD00043267
Crystalline/Powder; Molecular Formula: C8H13NO3·HCl; MW: 207.66
Melting Point: 170° (decomposes)
- Crystalline/Powder
Formula:
C₈H₁₃NO₃ MW: 207.65 g/mol Melting Pt: 170 °C (dec.) |
MDL Number:
MFCD00043267 CAS Number: 4644-61-5 EINECS: 225-073-8 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...