5-Bromo-2-formylpyridine ≥99%
Supplier: BeanTown Chemical
Synonyms:
5-Bromopyridine-2-carbaldehyde, 5-Bromopicolinaldehyde, 5-Bromopyridine-2-carboxaldehyde
215175-1G
215175-5G
BT215175-1GEA
88
USD
BT215175-1G
BT215175-5G
5-Bromo-2-formylpyridine ≥99%
5-Bromo-2-formylpyridine
CAS: 31181-90-5; MDL No: MFCD04112535
Solid; Molecular Formula: C6H4BrNO; MW: 186.01
Melting Point: 91-96°
Air Sensitive
- Air Sensitive
- Solid
Formula:
C₆H₄BrNO Melting Pt: 91…96 °C |
MDL Number:
MFCD04112535 CAS Number: 31181-90-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...