3-Amino-6-bromopyrazine-2-carboxylic acid ≥95%

Supplier: BeanTown Chemical

215170-1G 215170-5G
BT215170-1GEA 49 USD
BT215170-1G BT215170-5G
3-Amino-6-bromopyrazine-2-carboxylic acid ≥95%
3-Amino-6-bromopyrazine-2-carboxylic acid

CAS: 486424-37-7 Solid; Molecular Formula: C5H4BrN3O2; MW: 218.01


  • Solid

Formula: C₅H₄BrN₃O₂
MW: 218.01 g/mol
MDL Number: MFCD08275681
CAS Number: 486424-37-7

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR