Tributyl phosphate ≥98%

Supplier: BeanTown Chemical

Synonyms: TBP, TBPA

215165-1L 215165-250ML
BT215165-1LEA 72 USD
BT215165-1L BT215165-250ML
Tributyl phosphate ≥98%
Tributyl phosphate

CAS: 126-73-8; EC No: 204-800-2; MDL No: MFCD00009436; RTECS: TC7700000 Liquid; Linear Formula: [CH3(CH2)3O]3PO; Molecular Formula: C12H27O4P; MW: 266.32 Melting Point: -79°; Boiling Point: 180-183°/22 mmHg; Flash point: 145°C (293°F) Density (g/mL): 0.979; Refractive Index: 1.424


  • RTECS: TC7700000
  • Refractive index: 1.424
  • Liquid

Formula: (CH₃(CH₂)₃O)₃PO
MW: 266.32 g/mol
Boiling Pt: 289
Melting Pt: -79
Density: 0.979 g/mL
Flash Pt: 145 °C (293 °F)
MDL Number: MFCD00009436
CAS Number: 126-73-8
EINECS: 204-800-2
Merck Index: 13,09692
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb9a0e9a8476470c1c50ab98587c559fec16cd8d&vid=6bcd8ac2f1665c53ba2c7ace4f34ebb3d890b549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR