D-(+)-Tryptophan ≥99%
Supplier: BeanTown Chemical
Synonyms:
H-D-Trp-OH
215145-100G
215145-25G
215145-5G
BT215145-100GEA
334
USD
BT215145-100G
BT215145-25G
BT215145-5G
D-(+)-Tryptophan ≥99%
D-(+)-Tryptophan
CAS: 153-94-6; EC No: 205-819-9; MDL No: MFCD00005647; RTECS: YN6129000
Powder; Molecular Formula: C11H12N2O2; MW: 204.23
Melting Point: 282-285° (decomposes)
- RTECS: YN6129000
- Powder
Formula:
C₁₁H₁₂N₂O₂ MW: 204.23 g/mol Melting Pt: 282…285 °C (dec.) |
MDL Number:
MFCD00005647 CAS Number: 153-94-6 EINECS: 205-819-9 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...