Bathocuproin sulfonate disodium salt hydrate ≥97%

Supplier: BeanTown Chemical

Synonyms: Bathocuproindisulphonic disodium salt hydrate

215130-1G 215130-250MG 215130-5G
BT215130-1GEA 91.16 USD
BT215130-1G BT215130-250MG BT215130-5G
Bathocuproin sulfonate disodium salt hydrate ≥97%
Bathocuproin sulphonate disodium salt hydrate

CAS: 52698-84-7; EC No: 258-111-7; MDL No: MFCD03012947 Powder; Molecular Formula: C26H18N2Na2O6S2·xH2O; MW: 564.54 (anhydrous)


  • Powder

Formula: C₂₆H₁₈N₂Na₂O₆S₂·H₂O
MW: 564.55 g/mol (anhy)
MDL Number: MFCD03012947
CAS Number: 52698-84-7
EINECS: 258-111-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR