Sodium stannate trihydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Sodium tin(IV) oxide trihydrate, Disodium tin trioxide trihydrate

215120-100G 215120-2.5KG 215120-500G
BT215120-100GEA 53.53 USD
BT215120-100G BT215120-2.5KG BT215120-500G
Sodium stannate trihydrate ≥98%
Sodium stannate trihydrate

CAS: 12209-98-2; EC No: 235-030-5; MDL No: MFCD00150189 Powder; Linear Formula: Na2SnO3·3H2O; MW: 266.73


  • Powder

Formula: Na₂O₃Sn·3H₂O
MW: 266.73
MDL Number: MFCD00150189
CAS Number: 12209-98-2
EINECS: 235-030-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR