Potassium tetrafluoroborate ≥99%
Supplier: BeanTown Chemical
215115-2KG
215115-500G
BT215115-2KGEA
164
USD
BT215115-2KG
BT215115-500G
Potassium tetrafluoroborate ≥99%
Potassium tetrafluoroborate
CAS: 14075-53-7; EC No: 237-928-2; MDL No: MFCD00011395; RTECS: ED2800000
Powder; Linear Formula: KBF4; MW: 125.91
Melting Point: 530°
Density (g/mL): 2.505
Moisture Sensitive
- RTECS: ED2800000
- Moisture Sensitive
- Powder
Formula:
BF₄K Melting Pt: 530 °C Density: 2.505 g/mL |
MDL Number:
MFCD00011395 CAS Number: 14075-53-7 EINECS: 237-928-2 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...