Nickel (II) oxide, green

Supplier: BeanTown Chemical

Synonyms: Nickel monoxide, Nickel oxide, Bunsenite

215110-1KG 215110-250G
BT215110-1KGEA 248.43 USD
BT215110-1KG BT215110-250G
Nickel (II) oxide, green
Nickel (II) oxide

CAS: 1313-99-1; EC No: 215-215-7; MDL No: MFCD00011145; RTECS: QR8400000 -400 Mesh Powder; Molecular Formula: NiO; MW: 74.71 Melting Point: 1984° Density (g/mL): 6.67


  • RTECS: QR8400000
  • -400 Mesh Powder

Formula: NiO
MW: 74.69 g/mol
Melting Pt: 1984 °C
Density: 6.67 g/mL
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR