Gallium(III) sulfate hydrate ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
215105-10G
215105-50G
BT215105-10GEA
153.41
USD
BT215105-10G
BT215105-50G
Gallium(III) sulfate hydrate ≥99.999% (trace metals basis)
Gallium(III) sulfate hydrate
CAS: 13780-42-2; MDL No: MFCD00149725
Crystalline Aggregates; Linear Formula: Ga2(SO4)3·xH2O; MW: 427.62 (anhydrous)
Melting Point: 105-110°
Density (g/mL): 3.86
Hygroscopic
- Hygroscopic
- Crystalline Aggregates
Formula:
Ga₂(SO₄)₃·xH₂O (x≈18) Melting Pt: 105…110 °C Density: 3.86 g/mL |
MDL Number:
MFCD00149725 CAS Number: 13780-42-2 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...