2-(Boc-amino)-5-cyanopyridine ≥97%
Supplier: BeanTown Chemical
215045-1G
215045-5G
BT215045-1GEA
239
USD
BT215045-1G
BT215045-5G
2-(Boc-amino)-5-cyanopyridine ≥97%
2-(Boc-amino)-5-cyanopyridine
CAS: 814263-30-4; MDL No: MFCD07781163
Solid; Molecular Formula: C11H13N3O2 ; MW: 219.24
Melting Point: 170-175°
- Solid
Formula:
C₁₁H₁₃N₃O₂ Melting Pt: 170…175 °C |
MDL Number:
MFCD07781163 CAS Number: 814263-30-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...