Calcium acetate monohydrate ≥99.0% ACS
Supplier: BeanTown Chemical
214995-1KG
214995-250G
BT214995-1KGEA
204
USD
BT214995-1KG
BT214995-250G
Calcium acetate monohydrate ≥99.0% ACS
Calcium acetate monohydrate
CAS: 5743-26-0; EC No: 200-540-9; MDL No: MFCD00150019; RTECS: AF7875000
Powder; Linear Formula: Ca(OOCCH3)2·H2O; Molecular Formula: C4H6CaO4·H2O; MW: 176.18
Hygroscopic
- RTECS: AF7875000
- Hygroscopic
- Powder
Formula:
(H₃CCOO)₂Ca·1H₂O |
MDL Number:
MFCD00150019 CAS Number: 5743-26-0 EINECS: 200-540-9 |
Specification Test Results
Assay | 99.0% min. |
Insoluble matter | ≤0.005% |
Chloride (Cl) | ≤0.001% |
Sulfate (SO4) | ≤0.01% |
Barium (Ba) | ≤0.01% |
Potassium (K) | ≤0.01% |
Sodium (Na) | ≤0.02% |
Strontium (Sr) | ≤0.05% |
Heavy metals (as Pb) | ≤0.005% |
Iron (Fe) | ≤0.001% |
Magnesium (Mg) | ≤0.05% |
Titrable acid | ≤0.035 meq/g |
Alkalinity | Passes test |
Appearance | Colorless/white crystalline powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...