N,N-Dimethylglycine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid

214975-100G 214975-25G 214975-5G
BT214975-100GEA 710 USD
BT214975-100G BT214975-25G BT214975-5G
N,N-Dimethylglycine ≥98%
N,N-Dimethylglycine

CAS: 1118-68-9; EC No: 214-267-8; MDL No: MFCD00004283; RTECS: MB9865000 Crystalline/Powder; Linear Formula: (CH3)2NCH2COOH; Molecular Formula: C4H9NO2; MW: 103.12 Melting Point: 178-182° Hygroscopic


  • RTECS: MB9865000
  • Hygroscopic
  • Crystalline/Powder

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: 178…182 °C
MDL Number: MFCD00004283
CAS Number: 1118-68-9
EINECS: 214-267-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR