Sodium octanoate ≥96%

Supplier: BeanTown Chemical

Synonyms: Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate

214945-100G 214945-500G
BT214945-100GEA 47.78 USD
BT214945-100G BT214945-500G
Sodium octanoate ≥96%
Sodium octanoate

CAS: 1984-06-1; EC No: 217-850-5; MDL No: MFCD00058511; RTECS: RH0787000 Powder; Linear Formula: CH3(CH2)6COONa; Molecular Formula: C8H15NaO2; MW: 166.20


  • RTECS: RH0787000
  • Powder

Formula: C₈H₁₅NaO₂
MW: 166.2 g/mol
MDL Number: MFCD00058511
CAS Number: 1984-06-1
EINECS: 217-850-5

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR