5-Bromo-2-thenoic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Bromothiophene-2-carboxylic acid, 5-Bromo-2-thiophenecarboxylic acid

214935-1G 214935-25G 214935-5G
BT214935-1GEA 30 USD
BT214935-1G BT214935-25G BT214935-5G
5-Bromo-2-thenoic acid ≥98%
5-Bromo-2-thenoic acid

CAS: 7311-63-9; MDL No: MFCD00079725 Powder; Molecular Formula: C5H3BrO2S; MW: 207.05 Melting Point: 139-143°


  • Powder

Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
Melting Pt: 139…143 °C
MDL Number: MFCD00079725
CAS Number: 7311-63-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR