Triacetin (glycerol triacetate) ≥99%

Supplier: BeanTown Chemical

Synonyms: Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate

214920-2.5KG 214920-500G
BT214920-2.5KGEA 131.03 USD
BT214920-2.5KG BT214920-500G
Triacetin (glycerol triacetate) ≥99%
Triacetin (glycerol triacetate)

CAS: 102-76-1; EC No: 203-051-9; MDL No: MFCD00008716 Liquid; Molecular Formula: C9H14O6; MW: 218.21 Melting Point: 3°; Boiling Point: 258°; Flash point: 138°C (280°F) Density (g/mL): 1.16


  • Liquid

Formula: C₉H₁₄O₆
MW: 218.21 g/mol
Boiling Pt: 258 °C
Melting Pt: 3 °C
Density: 1.16 g/mL
Flash Pt: 138 °C (280 °F)
MDL Number: MFCD00008716
CAS Number: 102-76-1
EINECS: 203-051-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR