Palmitic acid ≥95%

Supplier: BeanTown Chemical

Synonyms: Hexadecanoic acid, Hexadecylic acid

214885-10KG 214885-2.5KG 214885-500G
BT214885-10KGEA 274 USD
BT214885-10KG BT214885-2.5KG BT214885-500G
Palmitic acid ≥95%
Palmitic acid

CAS: 57-10-3; EC No: 200-312-9; MDL No: MFCD00002747; RTECS: RT4550000 Flakes; Linear Formula: CH3(CH2)14COOH; Molecular Formula: C16H32O2; MW: 256.43 Melting Point: 62-65°; Boiling Point: 271.5°/100 mmHg; Flash point: 110°C (230°F) Density (g/mL): 0.852; Refractive Index: 1.427


  • RTECS: RT4550000
  • Refractive index: 1.427
  • Flakes

Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 271.5 °C/100 mmHg
Melting Pt: 62…65 °C
Density: 0.852 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR