Chloroquine diphosphate salt ≥98%
Supplier: BeanTown Chemical
Synonyms:
N4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate), N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate, Chloroquine phosphate
214870-100G
214870-25G
BT214870-100GEA
203.95
USD
BT214870-100G
BT214870-25G
Chloroquine diphosphate salt ≥98%
Chloroquine diphosphate salt
CAS: 50-63-5; EC No: 200-055-2; MDL No: MFCD00069852; RTECS: VB2450000
Solid; Molecular Formula: C18H26ClN3·2H3PO4; MW: 515.87
Melting Point: 200° (decomposes)
- RTECS: VB2450000
- Solid
Formula:
C₁₈H₂₆ClN₃ MW: 515.86 g/mol Melting Pt: 200 °C (dec.) |
MDL Number:
MFCD00069852 CAS Number: 50-63-5 EINECS: 200-055-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...