(S)-2-Methylpropane-2-sulfinamide ≥97%

Supplier: BeanTown Chemical

Synonyms: (S)-tert-Butylsulphinamide, (S)-(-)-tert-Butylsulphinamide, (S)-(-)-t-Butylsulphinamide, (S)-(-)-2-Methyl-2-propanesulphinamide, (S)-(-)-tert-Butyl sulphinamide

214865-1G 214865-25G 214865-5G
BT214865-1GEA 81 USD
BT214865-1G BT214865-25G BT214865-5G
(S)-2-Methylpropane-2-sulfinamide ≥97%
(S)-2-Methylpropane-2-sulfinamide

CAS: 343338-28-3; MDL No: MFCD05861480 Solid; Molecular Formula: C4H11NOS ; MW: 121.20 Melting Point: 97-101° Density (g/mL): 1.124; Optical Rotation: [α]20/D -4.5°, c = 1 in chloroform


  • Optical rotation: [α]20/D -4.5°, c = 1 in chloroform
  • Solid

Formula: C₄H₁₁NOS
MW: 121.20 g/mol
Melting Pt: 97…101 °C
Density: 1.124 g/mL
MDL Number: MFCD05861480
CAS Number: 343338-28-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR