4-Ethoxybenzaldehyde ≥97%
Supplier: BeanTown Chemical
CAS: 10031-82-0; EC No: 233-093-3; MDL No: MFCD00003388; RTECS: CU6100000
Liquid; Molecular Formula: C9H10O2; MW: 150.18
Melting Point: 13-14°; Boiling Point: 255°; Flash point:
- RTECS: CU6100000
- Refractive index: 1.559
- Air Sensitive
- Liquid
Formula:
C₉H₁₀O₂ Boiling Pt: 255 °C Melting Pt: 13…14 °C Density: 1.08 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00003388 CAS Number: 10031-82-0 EINECS: 233-093-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...