Dimethyl-2-nitroterephthalate ≥98%

Supplier: BeanTown Chemical

Synonyms: Dimethyl 2-nitroterephthalate

214730-1KG 214730-250G 214730-50G
BT214730-1KGEA 525.22 USD
BT214730-1KG BT214730-250G BT214730-50G
Dimethyl-2-nitroterephthalate ≥98%
Dimethyl-2-nitroterephthalate

CAS: 5292-45-5; EC No: 226-134-1; MDL No: MFCD00008422 Crystalline; Molecular Formula: C12H13NO6; MW: 239.18 Melting Point: 73-76° Density (g/mL): 1.272


  • Crystalline

Formula: C₁₀H₉NO₆
MW: 239.18 g/mol
Melting Pt: 73…76 °C
Density: 1.272 g/mL
MDL Number: MFCD00008422
CAS Number: 5292-45-5
EINECS: 226-134-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR