Rhenium ≥99.95% (trace metals basis), wire Ø 0.5 mm

Supplier: BeanTown Chemical

214715-100CM 214715-25CM 214715-5CM
BT214715-100CMEA 616 USD
BT214715-100CM BT214715-25CM BT214715-5CM
Rhenium ≥99.95% (trace metals basis), wire Ø 0.5 mm
Rhenium

CAS: 7440-15-5; EC No: 231-124-5; MDL No: MFCD00011195; RTECS: VI0780000 Solid; Molecular Formula: Re; MW: 186.21 Melting Point: 3180°; Boiling Point: 5596° Density (g/mL): 21.02


  • RTECS: VI0780000
  • Solid

Formula: Re
MW: 186.21 g/mol
Boiling Pt: 5596 °C
Melting Pt: 3180 °C
Density: 21.02 g/mL
MDL Number: MFCD00011195
CAS Number: 7440-15-5
EINECS: 231-124-5

Order Now

Specification Test Results

Assay 99.95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR