Rhodium(II) acetate dimer ≥98%

Supplier: BeanTown Chemical

Synonyms: Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)

214700-100MG 214700-2G 214700-500MG
BT214700-100MGEA 139 USD
BT214700-100MG BT214700-2G BT214700-500MG
Rhodium(II) acetate dimer ≥98%
Rhodium (II) acetate dimer

CAS: 15956-28-2; EC No: 240-084-8; MDL No: MFCD00003538; RTECS: VI9361000 Powder; Linear Formula: Rh2(OOCCH3)4; Molecular Formula: C8H12O8Rh2; MW: 441.99 Melting Point: 205°


  • RTECS: VI9361000
  • Powder

Formula: C₈H₁₂O₈Rh₂
MW: 441.987 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00003538
CAS Number: 15956-28-2
EINECS: 240-084-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR