Cytosine ≥98%

Supplier: BeanTown Chemical

214690-100G 214690-25G 214690-5G
BT214690-100GEA 550 USD
BT214690-100G BT214690-25G BT214690-5G
Cytosine ≥98%
Cytosine

CAS: 71-30-7; EC No: 200-749-5; MDL No: MFCD00006034; RTECS: UW7350150 Powder; Molecular Formula: C4H5N3O; MW: 111.10 Melting Point: 300°


  • RTECS: UW7350150
  • Powder

Formula: C₄H₅N₃O
Melting Pt: >300 °C
MDL Number: MFCD00006034
CAS Number: 71-30-7
EINECS: 200-749-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR