5-Bromo-2-methox-4-methylypyridine ≥96%

Supplier: BeanTown Chemical

Synonyms: 5-Bromo-2-methoxy-4-methylpyridine

214665-1G 214665-5G
BT214665-1GEA 60 USD
BT214665-1G BT214665-5G
5-Bromo-2-methox-4-methylypyridine ≥96%
5-Bromo-2-methox-4-methylypyridine

CAS: 164513-39-7; MDL No: MFCD04039980 Solid; Molecular Formula: C7H8BrNO; MW: 202.05 Melting Point: 33-37°; Flash point: 107°C (225°F)


  • Solid

Formula: C₇H₈BrNO
MW: 202.0485 g/mol
Melting Pt: 33…37 °C
Flash Pt: 107 °C (225 °F)
MDL Number: MFCD04039980
CAS Number: 164513-39-7

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR