Ytterbium(III) oxide ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Diytterbium trioxide, Ytterbium oxide

214655-10G 214655-50G
BT214655-10GEA 75 USD
BT214655-10G BT214655-50G
Ytterbium(III) oxide ≥99.99%
Ytterbium(III) oxide

CAS: 1314-37-0; EC No: 215-234-0; MDL No: MFCD00011290 Powder; Linear Formula: Yb2O3; MW: 394.08 Melting Point: 2227° Density (g/mL): 9.170 Air Sensitive, Hygroscopic


  • Air Sensitive, Hygroscopic
  • Powder

Formula: O₃Yb₂
MW: 394.08 g/mol
Melting Pt: 2346
Density: 9.170 g/mL
MDL Number: MFCD00011290
CAS Number: 1314-37-0
EINECS: 215-234-0

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR