2-Iodobiphenyl ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Iodo-1,1'-biphenyl
214650-1G
214650-5G
BT214650-1GEA
34
USD
BT214650-1G
BT214650-5G
2-Iodobiphenyl ≥98%
2-Iodobiphenyl
CAS: 2113-51-1; EC No: 218-303-3; MDL No: MFCD00039396; RTECS: DV5384800
Liquid; Molecular Formula: C12H9I; MW: 280.10
Boiling Point: 300°
Density (g/mL): 1.59; Refractive Index: 1.662
Light Sensitive
- RTECS: DV5384800
- Refractive index: 1.662
- Light Sensitive
- Liquid
Formula:
C₁₂H₉I MW: 280.11 g/mol Boiling Pt: 300 °C Density: 1.59 g/mL |
MDL Number:
MFCD00039396 CAS Number: 2113-51-1 EINECS: 218-303-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...