Lanthanum(III) oxide ≥99.99%
Supplier: BeanTown Chemical
214645-1KG
214645-250G
214645-50G
BT214645-1KGEA
691.11
USD
BT214645-1KG
BT214645-250G
BT214645-50G
Lanthanum(III) oxide ≥99.99%
Lanthanum(III) oxide
CAS: 1312-81-8; EC No: 215-200-5; MDL No: MFCD00011071
Powder; Molecular Formula: La2O3; MW: 325.82
Melting Point: 2307°
Density (g/mL): 6.51
Hygroscopic
- Hygroscopic
- Powder
Formula:
La₂O₃ MW: 325.81 g/mol Boiling Pt: 4200 Melting Pt: 2315 Density: 6.51 g/mL |
MDL Number:
MFCD00011071 CAS Number: 1312-81-8 EINECS: 215-200-5 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...