5-Methyl-1,3-cyclohexanedione ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Methylcyclohexane-1,3-dione

214640-1G 214640-25G 214640-5G
BT214640-1GEA 33 USD
BT214640-1G BT214640-25G BT214640-5G
5-Methyl-1,3-cyclohexanedione ≥98%
5-Methyl-1,3-cyclohexanedione

CAS: 4341-24-6; MDL No: MFCD00010379 Powder; Molecular Formula: C7H10O2; MW: 126.15 Melting Point: 126-130°


  • Powder

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Melting Pt: 126…130 °C
MDL Number: MFCD00010379
CAS Number: 4341-24-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR