(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol ≥98%
Supplier: BeanTown Chemical
Synonyms:
(1S,2S,3R,5S)-(+)-2,3-Pinanediol, (1S,2S,3R,5S)-(+)-Pinanediol
214605-1G
214605-25G
214605-5G
BT214605-1GEA
35
USD
BT214605-1G
BT214605-25G
BT214605-5G
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol ≥98%
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol
CAS: 18680-27-8; MDL No: MFCD00077851
Crystalline/Powder; Molecular Formula: C10H18O2; MW: 170.25
Melting Point: 57-59°; Flash point:
- Optical rotation: [α]20/D +8.5°, c = 6.5 in toluene
- Crystalline/Powder
Formula:
C₁₀H₁₈O₂ MW: 170.25 g/mol Melting Pt: 57…59 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00077851 CAS Number: 18680-27-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...