Potassium wolframate ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Potassium tungstate, Potassium tungsten oxide
214600-100G
214600-25G
BT214600-100GEA
180
USD
BT214600-100G
BT214600-25G
Potassium wolframate ≥99.5% (trace metals basis)
Potassium wolframate
CAS: 7790-60-5; EC No: 232-215-2; MDL No: MFCD00049668
-60 Mesh Powder; Linear Formula: K2WO4; MW: 326.04
Melting Point: 921°
Density (g/mL): 3.12
Hygroscopic
- Hygroscopic
- -60 Mesh Powder
Formula:
K₂O₄W MW: 326.03 g/mol Melting Pt: 921 °C Density: 3.12 g/mL |
MDL Number:
MFCD00049668 CAS Number: 7790-60-5 EINECS: 232-215-2 |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...