(S)-(-)-Hydroxypyrrolidine ≥99%, ee 99%
Supplier: BeanTown Chemical
Synonyms:
(S)-(-)-3-Pyrrolidinol, (S)-Pyrrolidin-3-ol, (S)-(-)-3-Hydroxypyrrolidine
214550-1G
214550-250MG
214550-5G
BT214550-1GEA
164
USD
BT214550-1G
BT214550-250MG
BT214550-5G
(S)-(-)-Hydroxypyrrolidine ≥99%, ee 99%
(S)-(-)-Hydroxypyrrolidine
CAS: 100243-39-8; MDL No: MFCD00192426
Liquid; Molecular Formula: C4H9NO; MW: 87.12
Flash point: 109°C (228°F)
Density (g/mL): 1.078; Refractive Index: 1.490; Optical Rotation: [α]20/D -6.5±1.5°, c = 3.5% in methanol
Air Sensitive, Hygroscopic
- Optical rotation: [α]20/D -6.5±1.5°, c = 3.5% in methanol
- Refractive index: 1.490
- Air Sensitive, Hygroscopic
- Liquid
Formula:
C₄H₉NO Density: 1.078 g/mL Flash Pt: 109 °C (228 °F) |
MDL Number:
MFCD00192426 CAS Number: 100243-39-8 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...