Tetrabutylammonium hydrogen sulfate ≥97%

Supplier: BeanTown Chemical

Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate

214530-1KG 214530-250G 214530-50G
BT214530-1KGEA 309 USD
BT214530-1KG BT214530-250G BT214530-50G
Tetrabutylammonium hydrogen sulfate ≥97%
Tetrabutylammonium hydrogen sulphate

CAS: 32503-27-8; EC No: 251-068-5; MDL No: MFCD00011637 Solid; Linear Formula: [CH3(CH2)3]4N(HSO4); Molecular Formula: C16H37NO4S; MW: 339.54 Melting Point: 169-171° Density (g/mL): 1.01 Hygroscopic


  • Hygroscopic
  • Solid

Formula: (C₄H₉)₄NHSO₄
MW: 339.54 g/mol
Melting Pt: 169…171 °C
Density: 1.01 g/mL
MDL Number: MFCD00011637
CAS Number: 32503-27-8
EINECS: 251-068-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR