Tungsten disulphide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Tungsten sulphide, Tungsten (IV) sulphide
214525-250G
214525-50G
BT214525-250GEA
261
USD
BT214525-250G
BT214525-50G
Tungsten disulphide ≥99.5%
Tungsten disulphide
CAS: 12138-09-9; EC No: 235-243-3; MDL No: MFCD00011467
Powder; Linear Formula: WS2; MW: 247.98
Melting Point: 1250° (decomposes)
Density (g/mL): 7.5
- Powder
Formula:
S₂W MW: 247.97 Melting Pt: 1250 °C (dec.) Density: 7.5 g/mL |
MDL Number:
MFCD00011467 CAS Number: 12138-09-9 EINECS: 235-243-3 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...