4-Methoxy-3-nitropyridine ≥98%
Supplier: BeanTown Chemical
214505-1G
214505-25G
214505-5G
BT214505-1GEA
54
USD
BT214505-1G
BT214505-25G
BT214505-5G
4-Methoxy-3-nitropyridine ≥98%
4-Methoxy-3-nitropyridine
CAS: 31872-62-5; MDL No: MFCD00209661
Solid; Molecular Formula: C6H6N2O3; MW: 154.12
Melting Point: 72-77°
- Solid
Formula:
C₆H₆N₂O₃ Melting Pt: 72…77 °C |
MDL Number:
MFCD00209661 CAS Number: 31872-62-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...