2-Amino-3-formylpyridine 98

Supplier: BeanTown Chemical

Synonyms: 2-Amino-3-pyridinecarboxaldehyde, 2-Aminonicotinaldehyde, 2-Aminopyridine-3-carboxaldehyde

214495-1G 214495-5G
BT214495-1GEA 66 USD
BT214495-1G BT214495-5G
2-Amino-3-formylpyridine 98
2-Amino-3-formylpyridine

CAS: 7521-41-7; MDL No: MFCD01830382 Solid; Molecular Formula: C6H6N2O ; MW: 122.12 Melting Point: 98-102° Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₆H₆N₂O
MW: 122.13
Melting Pt: 98…102 °C
MDL Number: MFCD01830382
CAS Number: 7521-41-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR