2-Amino-3-iodopyridine ≥98%
Supplier: BeanTown Chemical
214470-1G
214470-5G
BT214470-1GEA
142
USD
BT214470-1G
BT214470-5G
2-Amino-3-iodopyridine ≥98%
2-Amino-3-iodopyridine
CAS: 104830-06-0; MDL No: MFCD01651885
Solid; Molecular Formula: C5H5IN2 ; MW: 220.01
Melting Point: 87-91°
Air Sensitive, Light Sensitive
- Air Sensitive, Light Sensitive
- Solid
Formula:
C₅H₅IN₂ MW: 220.01 Melting Pt: 87…91 °C |
MDL Number:
MFCD01651885 CAS Number: 104830-06-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...