2-Amino-4-chloropyridine ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Chloro-2-pyridylamine, 4-Chloropyridin-2-amine

214455-25G 214455-5G
BT214455-25GEA 223 USD
BT214455-25G BT214455-5G
2-Amino-4-chloropyridine ≥97%
2-Amino-4-chloropyridine

CAS: 19798-80-2; MDL No: MFCD04113820 Crystalline; Molecular Formula: C5H6ClN2; MW: 128.56 Melting Point: 129-133°


  • Crystalline

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 129…133 °C
MDL Number: MFCD04113820
CAS Number: 19798-80-2

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR