Bicine ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine

214435-1KG 214435-250G 214435-50G
BT214435-1KGEA 443.19 USD
BT214435-1KG BT214435-250G BT214435-50G
Bicine ≥99%
Bicine

CAS: 150-25-4; EC No: 205-755-1; MDL No: MFCD00004295; RTECS: MB9700000 Powder; Molecular Formula: C6H13NO4; MW: 163.17


  • RTECS: MB9700000
  • Powder

Formula: C₆H₁₃NO₄
MW: 163.17 g/mol
MDL Number: MFCD00004295
CAS Number: 150-25-4
EINECS: 205-755-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR