(Acetylmethylene)triphenylphosphorane ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-(Triphenylphosphoranylidene)propan-2-one, 1-Triphenylphosphoranylidene-2-propanone

214420-10G 214420-250G 214420-50G
BT214420-10GEA 43 USD
BT214420-10G BT214420-250G BT214420-50G
(Acetylmethylene)triphenylphosphorane ≥99%
(Acetylmethylene)triphenylphosphorane

CAS: 1439-36-7; EC No: 215-878-2; MDL No: MFCD00008774; RTECS: UC3900000 Crystalline; Molecular Formula: C21H19OP; MW: 318.36 Melting Point: 203-205° Air Sensitive


  • RTECS: UC3900000
  • Air Sensitive
  • Crystalline

Formula: C₂₁H₁₉OP
MW: 318.36 g/mol
Melting Pt: 203…205 °C
MDL Number: MFCD00008774
CAS Number: 1439-36-7
EINECS: 215-878-2

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR