Kojic acid ≥99%
Supplier: BeanTown Chemical
214410-100G
214410-25G
214410-5G
BT214410-100GEA
572
USD
BT214410-100G
BT214410-25G
BT214410-5G
Kojic acid ≥99%
Kojic acid
CAS: 501-30-4; EC No: 207-922-4; MDL No: MFCD00006580; RTECS: UQ0875000
Crystalline/Powder; Molecular Formula: C6H6O4; MW: 142.11
Melting Point: 152-155°
- RTECS: UQ0875000
- Crystalline/Powder
Formula:
C₆H₆O₄ Melting Pt: 152…155 °C |
MDL Number:
MFCD00006580 CAS Number: 501-30-4 EINECS: 207-922-4 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...