Phenyl vinyl sulphone ≥99%
Supplier: BeanTown Chemical
Synonyms:
Vinyl phenyl sulphone, (Vinylsulphonyl)benzene
214370-1G
214370-25G
214370-5G
BT214370-1GEA
31
USD
BT214370-1G
BT214370-25G
BT214370-5G
Phenyl vinyl sulphone ≥99%
Phenyl vinyl sulphone
CAS: 5535-48-8; EC No: 226-890-2; MDL No: MFCD00007554
Crystalline; Linear Formula: C6H5SO2CH=CH2; Molecular Formula: C8H8O2S; MW: 168.22
Melting Point: 67-71°
- Crystalline
Formula:
C₈H₈O₂S Melting Pt: 67…71 °C |
MDL Number:
MFCD00007554 CAS Number: 5535-48-8 EINECS: 226-890-2 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...