Calcium acetate xhydrate ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
214350-25G
214350-5G
BT214350-25GEA
271
USD
BT214350-25G
BT214350-5G
Calcium acetate xhydrate ≥99.995% (trace metals basis)
Calcium acetate xhydrate
CAS: 114460-21-8; EC No: 200-540-9; MDL No: MFCD00012448; RTECS: AF7525000
Powder; Linear Formula: Ca(OOCCH3)2·xH2O; MW: 158.17 (anhydrous)
Hygroscopic
- RTECS: AF7525000
- Hygroscopic
- Powder
Formula:
(H₃CCOO)₂Ca·nH₂O MW: 176.18 g/mol |
MDL Number:
MFCD00150019 CAS Number: 114460-21-8 EINECS: 200-540-9 |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...